Ammonium heptamolybdate tetrahydrate analytical reagent
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate
04190.0600
04190.1600
KRAF04190.0600EA
0
GBP
KRAF04190.0600
KRAF04190.1600
Ammonium heptamolybdate tetrahydrate analytical reagent
Ammonium heptamolybdate tetrahydrate
Formula:
(NH₄)₆Mo₇O₂₄·4H₂O MW: 1235.86 g/mol Density: 2.498 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167059 CAS Number: 12054-85-2 EINECS: 234-722-4 Merck Index: 13,00536 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...