Sodium selenite pentahydrate
Supplier: ABCR
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AB118831.1000
AB118831.0250
ABCRAB118831.1000EA
0
GBP
ABCRAB118831.1000
ABCRAB118831.0250
Sodium selenite pentahydrate
Sodium selenite pentahydrate
Formula:
Na₂O₃Se·5H₂O MW: 263.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149173 CAS Number: 26970-82-1 EINECS: 233-267-9 UN: 2630 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...