Magnesium acetate tetrahydrate ≥99%, Reagent Grade

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Magnesium di(acetate) tetrahydrateAcetic acid magnesium salt tetrahydrate

M0631-1KGEA 219 GBP
M0631-1KG
Magnesium acetate tetrahydrate ≥99%, Reagent Grade
Magnesium acetate tetrahydrate

  • ≥95% for general laboratory use.

Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 68 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now

Specification Test Results

Assay 99 - 102 %
Insoluble in acetic acid Max. 0.005 %
Form solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR