Malachite green chloride
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N-(4-((4-(Dimethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride
, Basic Green 4
, N,N,N′,N′-Tetramethyl-4,4′-diaminotriphenylcarbenium chloride
, CI 42000
, Malachite green hydrochloride
, Malachite green
MOLEM81713410EA
0
GBP
MOLEM81713410
Malachite green chloride
Malachite green chloride
Formula:
C₂₃H₂₅ClN₂ MW: 364.92 g/mol |
MDL Number:
MFCD00011751 CAS Number: 569-64-2 EINECS: 209-322-8 UN: 2811 ADR: 6.1,III Merck Index: 12,05739 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...