Lithiumbis(trimethylsilyl)amide 24% in THF

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt

SILS09630.25 SILS09630.100
SILNSILS09630.25EA 0 GBP
SILNSILS09630.25 SILNSILS09630.100
Lithiumbis(trimethylsilyl)amide 24% in THF
Lithium bis(trimethylsilyl)amide
Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
MDL Number: MFCD00008261
CAS Number: 4039-32-1
EINECS: 223-725-6
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR