Glutaraldehyde 50% in aqueous solution
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Glutaral, Glutardialdehyde, 1,5-Pentanedial
EMR144010
EMR144025
EMR144100
EURLEMR144010EA
0
GBP
EURLEMR144010
EURLEMR144025
EURLEMR144100
Glutaraldehyde 50% in aqueous solution
Glutaraldehyde
Formula:
C₅H₈O₂ MW: 100.12 g/mol Density: 1.13 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007025 CAS Number: 111-30-8 EINECS: 203-856-5 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...