(±)-trans-1,2-Cyclohexanediol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(±)-trans-Cyclohexane-1,2-diol, trans-Cyclohexane-1,2-diol
008843-10G
008843-50G
FLUO008843-10GEA
0
GBP
FLUO008843-10G
FLUO008843-50G
(±)-trans-1,2-Cyclohexanediol
(±)-trans-1,2-Cyclohexanediol
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 237 °C (1013 hPa) Melting Pt: 100…103 °C Density: 1.147 g/cm³ (24 °C) |
MDL Number:
MFCD00063611 CAS Number: 1460-57-7 EINECS: 215-956-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...