DL-Valine

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (±)-ValineH-DL-Val-OH2-Amino-3-methylbutanoic acid

ROTH7540.1EA 60.2 GBP
ROTH7540.1
DL-Valine
DL-Valine
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.352 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS Number: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR