DL-Valine
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(±)-Valine
, H-DL-Val-OH
, 2-Amino-3-methylbutanoic acid
MOLE15070583-100GEA
0
GBP
MOLE15070583-100G
DL-Valine
DL-Valine
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Boiling Pt: 217 °C (1013 hPa) Melting Pt: 86 °C Density: 1.352 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004267 CAS Number: 516-06-3 EINECS: 208-220-0 Merck Index: 12,10046 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...