(±)-2-Nonanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (±)-Heptyl methyl carbinol, (±)-2-Hydroxy nonane, (±)-Methyl heptyl carbinol

M-Clarity™ quality level = MQ300

W331503-100G W331503-1KG W331503-SAMPLE
SIALW331503-100GEA 185 GBP
SIALW331503-100G SIALW331503-1KG SIALW331503-SAMPLE
(±)-2-Nonanol, Sigma-Aldrich®
(±)-2-Nonanol
Formula: C₉H₂₀O
MW: 144.26 g/mol
Boiling Pt: 193 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.831 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004593
CAS Number: 628-99-9
EINECS: 211-065-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR