DL-Lithium lactate
Supplier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(±)-2-Hydroxypropanoic acid lithium salt
, DL-Lactic acid lithium salt
, (±)-Lactic acid lithium salt
, Lithium 2-hydroxypropanoate
, (±)-2-Hydroxypropionic acid lithium salt
PROOBP433EA
443
GBP
PROOBP433
DL-Lithium lactate
DL-Lithium lactate
Formula:
H₃CCH(OH)COOLi MW: 96.01 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065399 CAS Number: 867-55-0 EINECS: 212-761-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...