(±)-2-Bromopropionic acid
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
DL-2-Bromopropanoic acid
, 2-Bromopropanoic acid
, DL-2-Bromopropionic acid
, (±)-2-Bromopropanoic acid
EHERC10760700EA
107
GBP
EHERC10760700
(±)-2-Bromopropionic acid
(±)-2-Bromopropionic acid
Formula:
H₃CBrCHCOOH MW: 152.98 g/mol Boiling Pt: 203 °C (1013 hPa) Melting Pt: ∼25 °C Density: 1.7 g/cm³ (25 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00004211 CAS Number: 598-72-1 EINECS: 209-947-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...