Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

EMR193001 EMR193500
EURLEMR193001EA 0 GBP
EURLEMR193001 EURLEMR193500
Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90)
Polyvinylpyrrolidone
Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR