Polyethylene glycol 8,000, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: PEG 8000Ethylene glycol polymer 8000PEG 8,000

M-Clarity™ quality level = MQ100

81294-1GEA 184 GBP
81294-1G
Polyethylene glycol 8,000, Supelco®
Polyethylene glycol 8,000
Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Boiling Pt: 250 °C (1013 hPa)
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR