Polyethylene glycol 300, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: PEG 300

M-Clarity™ quality level = MQ100

81283-1GEA 176 GBP
81283-1G
Polyethylene glycol 300, Supelco®
Polyethylene glycol 300
Formula: H(OCH₂CH₂)nOH
MW: 300 g/mol
Melting Pt: –15…–10 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR