Cellulose, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Microcrystallinecellulose

M-Clarity™ quality level = MQ100

22184-250G-FEA 95.4 GBP
22184-250G-F
Cellulose, Supelco®
Cellulose
Formula: (C₁₂H₂₂O₁₁)n
MW: 342.3 g/mol
Melting Pt: 260…270 °C
Density: 1.27…1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081512
CAS Number: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR