Cellulose, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Microcrystallinecellulose
M-Clarity™ quality level = MQ100
09906-1KGEA
183
GBP
09906-1KG
Cellulose, Supelco®
Cellulose
Formula:
(C₁₂H₂₂O₁₁)n MW: 342.3 g/mol Melting Pt: 260…270 °C Density: 1.27…1.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00081512 CAS Number: 9004-34-6 EINECS: 232-674-9 Merck Index: 13,01977 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...