D-(-)-Arabinose, Millipore®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal
M-Clarity™ quality level = MQ100
10850-100G
10850-25G
SIAL10850-100GEA
192
GBP
SIAL10850-100G
SIAL10850-25G
D-(-)-Arabinose, Millipore®
D(-)-Arabinose
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Melting Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135608 CAS Number: 10323-20-3 EINECS: 233-708-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...