trans-Platin
Supplier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
trans-Dichlorodiamineplatinum(II)
, trans-Diamminedichloroplatinum (II)
, trans-Diammineplatinum (II) dichloride
, trans-Dichlorodiammine platinum(II)
PROOBP544EA
380
GBP
PROOBP544
trans-Platin
trans-Platin
Formula:
H₆Cl₂N₂Pt MW: 300.05 g/mol Melting Pt: 340 °C Density: 3,746 g/cm³ (20 °C) |
MDL Number:
MFCD00011623 CAS Number: 14913-33-8 EINECS: 238-980-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...