Decacarbonyldirhenium, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Rhenium carbonyl

M-Clarity™ quality level = MQ100

SIAL245003-5GEA 712 GBP
SIAL245003-5G
Decacarbonyldirhenium, Sigma-Aldrich®
Decacarbonyldirhenium
Formula: C₁₀O₁₀Re₂
MW: 652.52 g/mol
Melting Pt: 181…184 °C
Density: 2.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011198
CAS Number: 14285-68-8
EINECS: 238-202-8
UN: 3466
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR