Molecular sieve 3A (0.3 nm, 3 Å), beads, TECHNICAL, Ø 1.6 - 2.5 mm
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Linde 3A
, Molecular sieve 0.3 nm
, Sodium aluminium silicate
, Molecular sieve A3
, 3A molecular sieve
28463.292EA
184
GBP
28463.292
Molecular sieve 3A (0.3 nm, 3 Å), beads, TECHNICAL, Ø 1.6 - 2.5 mm
Molecular sieve 3A (0.3 nm, 3 Å)
Adsorption of molecules smaller than 3 µm in diameter
Formula:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00131613 CAS Number: 308080-99-1 EINECS: 215-283-8 |
Specification Test Results
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...