Cobalt, powder, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
1.12211.0100EA
105
GBP
1.12211.0100
1.12211.9999
Cobalt, powder, Sigma-Aldrich®
Cobalt
Formula:
Co MW: 58.93 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 ADR: 4.1,II |
Specification Test Results
Solubility | (20 °C) practically insoluble |
Melting Point | 1493 °C |
Molar Mass | 58.93 g/mol |
Bulk Density | 850 kg/m3 |
Boiling Point | 2870 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...