Rhenium, powder, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL267279-5GEA
560
GBP
SIAL267279-5G
SIAL267279-25G
Rhenium, powder, Sigma-Aldrich®
Rhenium
Formula:
Re MW: 186.21 g/mol |
MDL Number:
MFCD00011195 CAS Number: 7440-15-5 EINECS: 231-124-5 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...