Rhenium, powder, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

SIAL204188-5GEA 619 GBP
SIAL204188-5G
Rhenium, powder, Sigma-Aldrich®
Rhenium
Formula: Re
MW: 186.21 g/mol
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR