Rhenium, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

SIAL459984-13GEA 1180 GBP
SIAL459984-13G
Rhenium, Sigma-Aldrich®
Rhenium
Formula: Re
MW: 186.21 g/mol
Boiling Pt: 5630 °C (1013 hPa)
Melting Pt: 3180 °C
Density: 21.02 g/cm³ (20 °C)
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
Merck Index: 12,08343

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR