4-Formyl-3-hydroxybenzoic acid ≥96.5% (by GC), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

633461-1G 633461-5G
SIAL633461-1GEA 168 GBP
SIAL633461-1G SIAL633461-5G
4-Formyl-3-hydroxybenzoic acid ≥96.5% (by GC), Sigma-Aldrich®
4-Formyl-3-hydroxybenzoic acid
Formula: C₈H₆O₄
MW: 166.1308 g/mol
Melting Pt: 235…240 °C (lit.)
MDL Number: MFCD05865188
CAS Number: 619-12-5

Order Now

Specification Test Results

Infrared spectrum Conforms to structure
Purity (titration by NaOH) 96.5 - 103.5%
GC (area%) ≥96.5%
Appearance (colour) White to Dark Yellow and Faint Beige to
Dark Beige and Faint Brown to Dark Brown

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR