Bromine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

SIAL470864-50GEA 246 GBP
SIAL470864-50G
Bromine, Sigma-Aldrich®
Bromine
Formula: Br₂
MW: 159.81 g/mol
Boiling Pt: 58.7 °C (1013 hPa)
Melting Pt: –7.2 °C
Density: 3.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010896
CAS Number: 7726-95-6
EINECS: 231-778-1
UN: 1744
ADR: 8,I
Merck Index: 13,01378

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR