Silicon ≥98% (metals basis), crystalline powder -140 mesh

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

39688.18EA 79.5 GBP
39688.18 39688.30
Silicon ≥98% (metals basis), crystalline powder -140 mesh
Silicon

For plasma spraying

Formula: Si
MW: 28.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR