Dissection board, cork
Supplier: CellPath
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CLLPCHF-0100-00AEA 84.6 GBP
CLLPCHF-0100-00A
Dissection board, cork
Dissecting Boards
This cork dissection board is resistant to absorption and permits thorough rinsing.
  • Ideal for pinning out samples
  • Can be written on to illustrate tumour locations which is especially useful for cervix or neck disections