Glove box, hypoxia chamber, 856-HYPO
Supplier: Plas-Labs
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
135-0690EA 0 GBP
135-0690
Glove box, hypoxia chamber, 856-HYPO
Glove Boxes and Glove Bags
This new hypoxia chamber glove box is ideal for tissue culture work, including tumour cell and stem cell research. It features a colour touch screen for automatically controlling oxygen, temperature and carbon dioxide levels. Ethernet port connection for data logging.
  • O₂ and CO₂ sensor
  • Password protected, configurable PID control
  • Platinum RTD with sealing gland
  • Air circulation fan
  • Inert gas hook-up
Configurable for up to six individually controlled sampling chambers.
  • O₂ and CO₂ sensor
  • Password protected, configurable PID control
  • Platinum RTD with sealing gland
  • Air circulation fan
  • Inert gas hook-up
Configurable for up to six individually controlled sampling chambers.