AMPIGENE® qPCR Probe Mix Hi-ROX
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENZNUC1060200 ENZNUC1061000
ENZOENZNUC1060200EA 136 GBP
ENZOENZNUC1060200 ENZOENZNUC1061000
AMPIGENE® qPCR Probe Mix Hi-ROX
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Optimised probe-based qPCR.
AMPIGENE® qPCR Probe Mixes use the latest developments in polymerase technology and buffer chemistry to deliver high performing probe-based real-time PCR with minimal optimization. AMPIGENE® qPCR Probe Mixes are designed for use on a variety of probes including: Taqman ®, Molecular Beacons ®, and Scorpion probes ®. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR Probe Mixes come with either Lo-ROX, Hi-ROX, or No-ROX concentration. For Lo-ROX and No-ROX, see Prod. Nos. ENZ-NUC105 and ENZ-NUC107. Choose the appropriate ROX concentration for the instrument.
Ordering information: Contains 2x AMPIGENE® qPCR Probe Mix Hi-ROX (includes dNTPs and polymerases).