PGE2 CLIA kit
Supplier: Enzo Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALEXADI-910-001EA 592 GBP
ALEXADI-910-001 ENZOADI910001
PGE2 CLIA kit
Assays ELISAs
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species.
The PGE2 CLIA kit is a sensitive chemiluminescent competitive enzyme immunoassay kit for the quantitative determination of PGE2 in biological fluids from any species.
Ordering information: Contains White GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, CLIA Substrate.