IFN-gamma (human), ELISA kit
Supplier: Enzo Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALEXADI-900-136EA 819 GBP
ALEXADI-900-136 ENZOADI900136
IFN-gamma (human), ELISA kit
Assays ELISAs
The IFN-gamma (Human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm.
  • Ultrasensitive
  • Efficient - only 50 µL diluted sample needed per well
  • Measure natural and recombinant human IFN-γ
For the quantitative determination of human IFN-gamma in culture supernatants, plasma, serum, and urine.
Delivery information: Supplied with microtiter plate, SA-HRP concentrate, SA-HRP dilution buffer, biotinylated antibody, standard, standard diluent, wash buffer concentrate, TMB substrate, stop solution