Anti-Y Mouse Monoclonal Antibody (Biotin) [clone: ]

Supplier: Enzo Life Sciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ALEXBML-SA243-0100EA 320 GBP
ALEXBML-SA243-0100 ENZOBMLSA2430100
Anti-Y Mouse Monoclonal Antibody (Biotin) [clone: ]
Antibodies
Host: Mouse, Isotype: IgG2b

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry

Recognizes phosphotyrosine. Species independent. No cross-reactivity with phosphoserine or phosphothreonine.

Type: Primary
Antigen: Y
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Many Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR