Anti-HLA-G Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ALEXALX805704C100EA 393 GBP
ALEXALX805704C100 ENZOALX805704C100
Anti-HLA-G Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Western Blot

Recognizes human denatured HLA-G heavy chain.

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR