Anti-DCC Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ALEXALX804853C100EA 527 GBP
ALEXALX804853C100 ENZOALX804853C100
Anti-DCC Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a lambda

Recognizes human DCC.

Type: Primary
Antigen: DCC
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR