Anti-TLR8 Mouse Monoclonal Antibody (Biotin)

Supplier: Enzo Life Sciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ALEXALX804376BC100EA 581 GBP
ALEXALX804376BC100 ENZOALX804376BC100
Anti-TLR8 Mouse Monoclonal Antibody (Biotin)
Antibodies
The Toll-like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules. The TRL family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition. Ten human homologs of TLRs (TLR1-10) have been described. TLR8 gene contains three exons, two of which have coding function. TLR8 cDNA codes for a protein of approximate molecular weight of 120kDa.

  • For ELISA, Flow Cytometry

Recognizes human and mouse TLR8. Detects a band of ~120kDa by Western blot.

Type: Primary
Antigen: TLR8
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR