Hydraulic presses, E-Z Press™
Supplier: International Crystal Laboratories
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
634-0080EA 9860 GBP
634-0080 INCR0012-6306
Hydraulic presses, E-Z Press™
Presses
E-Z Press™ presses handle applications including pressing KBr pellets for FTIR spectroscopy and briquetting samples for XRF spectroscopy.
  • PC safety shields with solid brass hardware
  • Die centering depressions in the round bottom platens
Ports in the walls allow access for vacuum lines and electrical cords, enabling use with evacuable dies and heated platens.
Delivery information: Supplied with a stainless steel filling disc.