Sodium hypochlorite 1.3 - 1.6% available chlorine, J.T.Baker®
Supplier: Avantor
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3941.0100PEEA
0
GBP
3941.0100PE
3941.1000PE
Sodium hypochlorite 1.3 - 1.6% available chlorine, J.T.Baker®
Sodium hypochlorite
Formula:
ClNaO MW: 74.44 g/mol |
MDL Number:
MFCD00011120 CAS Number: 7681-52-9 UN: 3212 ADR: 5.1,II |
Specification Test Results
Active Chlorine | 1.3 - 1.6 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...