Disinfectant, high level medical surface disinfectant, CHEMGENE HLD₄H

Supplier: Medimark Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

DIUKXTM302EA 86.4 GBP
DIUKXTM302
Disinfectant, high level medical surface disinfectant, CHEMGENE HLD₄H
Cleaning Agents Disinfectants
For bio-hazard contamination (blood, urine, vomit etc) dilute to 1:20 (5%). Ready-to-use solution can be prepared by diluting 50 ml of concentrate product in 1 litre of water of a suitable quality.

  • Ideal to wash down all surfaces with disinfectant at 1:50 (2%) and dry with paper towel or allow to dry naturally
  • Wipes can also be used to wipe surfaces and be allowed to dry

Caution: Use biocides safely. Always read the label and product information before use.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR