Aldehyde dehydrogenase

Supplier: Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


126925-100EA 146 GBP
126925-100
Aldehyde dehydrogenase
Enzymes
Aldehyde dehydrogenase is a native aldehyde dehydrogenase from yeast that catalyses the oxidation of acetaldehyde to acetic acid. Used for the determination of acetaldehyde.

  • Form: White lyophilised solid
  • Specific activity: ≥20 units/mg protein
  • Storage temperature: 2 to 8 °C

One unit is defined as the amount of enzyme that will oxidise 1,0 µmol of acetaldehyde to acetic acid per min at 25 °C, pH 8,0.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR