Syrup bottles, narrow neck
Supplier: APG Europe
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALUG1000019EA 44.4 GBP
ALUG1000019
Syrup bottles, narrow neck
Bottles Glass Bottles
These ready to fill syrup bottles are made of type 3 clear glass with screw neck PP 28.
  • Pharmaceutical grade moulded glass
  • SGD mold reference 52473
  • Light transmission for amber glass is full compliance with USP and EP
  • Clean, particulate and microbiological control
  • Type: alpha