Semi-electronic rotary microtome, 3005 E
Supplier: PFM Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-2973EA 26860 GBP
720-2973
Semi-electronic rotary microtome, 3005 E
Microtomes and Microtome Assemblies Microtomes
High performance manual rotary microtome.
  • Backlash- and maintenance-free horizontal and vertical cross-roller bearing system
  • Smooth-running handwheel
  • Handwheel locking system in each position
  • Quick change system for object clamps
  • Ease of object orientation with memory function
  • Section counter, section thickness addition and finger guard
Suitable for all applications in routine use, research and industry.
Ordering information: Includes a disposable blade holder, low profile (narrow blade), operating manual, section waste tray, and protective cover.