Anti-BrdU Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-0917R-CY5EA 805 GBP
BOSSBS-0917R-CY5
Anti-BrdU Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Bromodeoxyuridine is a synthetic nucleoside that is an analogue of thymidine. BrdU is incorporated into DNA during S-phase of a cell cycle. It is commonly used in the detection of proliferating cells in living tissues

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BrdU
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR