Anti-Snake poison Protein Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-0763R-FITCEA 805 GBP
BOSSBS-0763R-FITC
Anti-Snake poison Protein Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Snake poison Protein

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Snake poison Protein
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR