Anti-2,4-D Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-0861R-CY5.5EA 805 GBP
BOSSBS-0861R-CY5.5
Anti-2,4-D Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
2,4-Dichlorophenoxyacetic acid (2,4-D) is a common systemic herbicide used in the control of broadleaf weeds. It is the most widely used herbicide in the world, and the third most commonly used in North America. [1] 2,4-D is also an important synthetic auxin, often used in laboratories for plant research and as a supplement in plant cell culture media such as MS medium. 2,4-D is a synthetic auxin, which is a class of plant growth regulators. It is absorbed through the leaves and is translocated to the meristems of the plant. Uncontrolled, unsustainable growth ensues causing stem curl-over, leaf withering, and eventual plant death. 2,4-D is typically applied as an amine salt, but more potent ester versions exist as well.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: 2,4-D
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR