Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-2197R-A350EA 805 GBP
BOSSBS-2197R-A350
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
duck hepatitis A virus genome polyprotein

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: duck hepatitis A virus 1 Genome polyprotein
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR