Anti-Glucoamylase Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-13383R-CY7EA 805 GBP
BOSSBS-13383R-CY7
Anti-Glucoamylase Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Glucoamylase is an important industrial enzyme used in saccharification steps in both starch enzymatic conversion and in alcohol production. The catalytic domain degrades oligosaccharides from the non reducing end, releasing glucose, and the starch domain binds the enzyme to raw starch and to the cell wall.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glucoamylase
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR