Anti-APOBEC2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-12493R-CY3EA 805 GBP
BOSSBS-12493R-CY3
Anti-APOBEC2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
APOBEC2 is a 224 amino acid protein that belongs to the cytidine and deoxycytidylate deaminase family. Expressed exclusively in heart and skeletal muscle, APOBEC2 is thought to be a probable C (cytidine) to U (uridine) editing enzyme. However, unlike other members of the family, such as APOBEC1, which mediates the editing of apolipoprotein (apo) B mRNA, APOBEC2 does not display any detectable apoB mRNA editing activity. Also, APOBEC2 has been shown to have low, but definite, intrinsic cytidine deaminase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOBEC2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR