Anti-Sauvagine Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-11424R-CY3EA 805 GBP
BOSSBS-11424R-CY3
Anti-Sauvagine Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Sauvagine is a 40-residue peptide isolated and characterized from frog skin. It shares significant structural homology with and elicits similar biological responses as corticotropin-releasing factor (CRF). It has high affinity for both the CRF1 and CRF2 receptors (and low affinity for the CRF binding protein), suggesting its potential as a pharmacological tool for the study of these receptor sub-types. Due to the binding to CRF2 receptors and their role in the release of ACTH and beta-endorphin during stress, possible areas of research with this ligand include anxiety, depression and obesity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Sauvagine
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR